Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GOSR2 SIRNA Assays ›
Gene Symbol:
GOSR2
Gene Name:
golgi SNAP receptor complex member 2
Gene Aliases:
Bos1, EPM6, GS27
Chromosome Location:
Chr.17: 46923086 - 46975519 on Build GRCh38
UniGene ID:
Hs.463278
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
112309
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GOSR2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001012511.2
NP_001012529.1
4
297
NM_001321133.1
NP_001308062.1
4
297
NM_001321134.1
NP_001308063.1
4
345
NM_004287.4
NP_004278.2
4
297
NM_054022.3
NP_473363.1
4
297
XM_005257844.3
XP_005257901.1
4
306
XM_006722190.3
XP_006722253.1
4
306
XM_011525501.2
XP_011523803.1
4
306
XM_011525502.2
XP_011523804.1
4
306
XM_017025378.1
XP_016880867.1
4
303
XM_017025379.1
XP_016880868.1
4
297
XM_017025380.1
XP_016880869.1
4
654
XM_017025381.1
XP_016880870.1
4
280
XM_017025382.1
XP_016880871.1
4
632
XM_017025383.1
XP_016880872.1
4
306
XM_017025384.1
XP_016880873.1
4
328
XM_017025385.1
XP_016880874.1
4
303
XM_017025386.1
XP_016880875.1
4
306
XM_017025387.1
XP_016880876.1
4
303
XM_017025388.1
XP_016880877.1
4
306
XM_017025389.1
XP_016880878.1
4
313
XM_017025390.1
XP_016880879.1
4
658
XM_017025391.1
XP_016880880.1
4
656
XM_017025392.1
XP_016880881.1
4
306
XM_017025393.1
XP_016880882.1
4
303
XM_017025394.1
XP_016880883.1
4
654
XM_017025395.1
XP_016880884.1
4
654
GenBank mRNA
AF007548.1
-
4
221
AF229796.1
-
4
252
AK290890.1
-
4
272
BC009710.2
-
4
268
3890308
BC034762.1
-
4
240
5185659
BG718145.1
-
4
291
BM846811.1
-
4
291
CD048223.1
-
4
322
Back To Top
More Information
Panther Classification:
Molecular Function -
SNARE protein
Gene Ontology Categories:
Function(s)
Process(es)
protein targeting to vacuole
ER to Golgi vesicle-mediated transport
intra-Golgi vesicle-mediated transport
Golgi to vacuole transport
IRE1-mediated unfolded protein response
retrograde transport, endosome to Golgi
COPII vesicle coating
vesicle fusion with Golgi apparatus
membrane fusion
SNARE binding
transporter activity
SNAP receptor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline