Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GALT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080605.3 | NP_542172.2 | 1 | 1530 | ||
GenBank mRNA | BG290330.1 | - | 175 |