Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TLR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003266.3 | NP_003257.1 | 4 | 950 | ||
NM_138554.4 | NP_612564.1 | 3 | 830 | |||
NM_138557.2 | NP_612567.1 | 2 | 663 | |||
GenBank mRNA | AB445638.1 | - | 3 | 532 | ||
AK226155.1 | - | 4 | 705 | |||
AK290053.1 | - | 4 | 933 | |||
AK293068.1 | - | 3 | 661 | |||
AK303730.1 | - | 2 | 494 | |||
BC025294.1 | - | 2 | 449 | |||
BC117422.1 | - | 3 | 552 | 40125973 | ||
BC143841.1 | - | 4 | 672 | |||
U88880.1 | - | 4 | 696 | |||
U93091.1 | - | 3 | 635 |