Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144563.2 | NP_653164.2 | 9 | 1057 | ||
GenBank mRNA | AY050633.1 | - | 9 | 1033 | ||
BC015529.1 | - | 9 | 1029 | 3904718 | ||
L35035.1 | - | 4 | 461 |