- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: GCNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001491.2 | NP_001482.1 | 3 | 1795 | ||
NM_145649.4 | NP_663624.1 | 5 | 1644 | |||
NM_145655.3 | NP_663630.2 | 3 | 1323 | |||
XM_005248999.2 | XP_005249056.1 | 6 | 1442 | |||
XM_006715052.3 | XP_006715115.1 | 5 | 1721 | |||
GenBank mRNA | AB078432.1 | - | 3 | 1090 | ||
AB078433.1 | - | 3 | 1093 | |||
AF401652.1 | - | 3 | 1204 | |||
AF401653.1 | - | 3 | 1204 | |||
AF458024.1 | - | 3 | 1093 | |||
AF458025.1 | - | 3 | 1087 | |||
AF458026.1 | - | 3 | 1093 | |||
AF458027.1 | - | 3 | 1093 | |||
AF458028.1 | - | 3 | 1093 | |||
AF458029.1 | - | 3 | 1093 | |||
AF458030.1 | - | 3 | 1093 | |||
AK090483.1 | - | 3 | 1649 | |||
AK098105.1 | - | 3 | 487 | |||
AK291767.1 | - | 5 | 1628 | |||
AK307329.1 | - | 4 | 451 | |||
AK307337.1 | - | 5 | 524 | |||
AK307373.1 | - | 5 | 652 | |||
AK313426.1 | - | 3 | 1338 | |||
AK313903.1 | - | 3 | 1504 | |||
AL832714.1 | - | 3 | 346 | |||
AL832719.1 | - | 3 | 1324 | |||
AY435145.1 | - | 3 | 1093 | |||
AY435146.1 | - | 3 | 1087 | |||
AY435147.1 | - | 3 | 1093 | |||
BC074801.2 | - | 3 | 1107 | 30915526 | ||
BC074802.2 | - | 3 | 1107 | 30915312 | ||
BC130524.1 | - | 3 | 1265 | 40146555 | ||
BX647576.1 | - | 3 | 1322 | |||
BX647668.1 | - | 3 | 1324 | |||
L19659.1 | - | 3 | 1341 | |||
Z19550.1 | - | 3 | 1341 |