Gene Symbol: GCNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001491.2 | NP_001482.1 | 3 | 1920 | ||
NM_145649.4 | NP_663624.1 | 5 | 1769 | |||
NM_145655.3 | NP_663630.2 | 3 | 1448 | |||
XM_005248999.2 | XP_005249056.1 | 6 | 1567 | |||
XM_006715052.3 | XP_006715115.1 | 5 | 1846 | |||
GenBank mRNA | AF401652.1 | - | 3 | 1329 | ||
AF401653.1 | - | 3 | 1329 | |||
AK090483.1 | - | 3 | 1774 | |||
AK098105.1 | - | 3 | 612 | |||
AK291195.1 | - | 3 | 1504 | |||
AK291767.1 | - | 5 | 1753 | |||
AK307329.1 | - | 4 | 576 | |||
AK307337.1 | - | 5 | 649 | |||
AK307373.1 | - | 5 | 777 | |||
AL832714.1 | - | 3 | 471 | |||
AL832719.1 | - | 3 | 1449 | |||
AY435145.1 | - | 3 | 1218 | |||
AY435146.1 | - | 3 | 1212 | |||
AY435147.1 | - | 3 | 1218 | |||
BC074801.2 | - | 3 | 1232 | 30915526 | ||
BC074802.2 | - | 3 | 1232 | 30915312 | ||
BC130524.1 | - | 3 | 1390 | 40146555 | ||
BX647576.1 | - | 3 | 1447 | |||
BX647668.1 | - | 3 | 1450 | |||
L19659.1 | - | 3 | 1466 | |||
Z19550.1 | - | 3 | 1466 |