Gene Symbol: GCNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001491.2 | NP_001482.1 | 3 | 3134 | ||
NM_145649.4 | NP_663624.1 | 5 | 2983 | |||
NM_145655.3 | NP_663630.2 | 3 | 2662 | |||
XM_005248999.2 | XP_005249056.1 | 6 | 2781 | |||
XM_006715052.3 | XP_006715115.1 | 5 | 3060 | |||
GenBank mRNA | AK090483.1 | - | 3 | 2988 | ||
AK098105.1 | - | 3 | 1826 | |||
AL832714.1 | - | 3 | 1685 | |||
AL832719.1 | - | 3 | 2663 | |||
AY435145.1 | - | 3 | 2432 | |||
AY435146.1 | - | 3 | 2426 | |||
AY435147.1 | - | 3 | 2432 | |||
BC130524.1 | - | 3 | 2604 | 40146555 | ||
BX647576.1 | - | 3 | 2661 | |||
BX647668.1 | - | 3 | 2663 |