Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS6ST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077188.1 | NP_001070656.1 | 6 | 1863 | ||
NM_147175.3 | NP_671704.3 | 4 | 1743 | |||
XM_005262490.3 | XP_005262547.1 | 5 | 1763 | |||
XM_005262491.2 | XP_005262548.1 | 4 | 1676 | |||
XM_011531406.1 | XP_011529708.1 | 5 | 1092 | |||
XM_017029944.1 | XP_016885433.1 | 3 | 1643 | |||
XM_017029945.1 | XP_016885434.1 | 5 | 1037 | |||
XM_017029946.1 | XP_016885435.1 | 4 | 1001 | |||
GenBank mRNA | AB067776.1 | - | 4 | 1023 | ||
AB067777.1 | - | 2 | 890 | |||
AK027720.1 | - | 3 | 1019 | |||
AK056706.1 | - | 4 | 1743 | |||
AK075402.1 | - | 3 | 922 | |||
AK295041.1 | - | 4 | 1038 | |||
AK295118.1 | - | 4 | 1003 | |||
AK298956.1 | - | 2 | 547 | |||
AL049679.1 | - | 4 | 1004 | |||
BC037325.2 | - | 4 | 922 | |||
BC094718.1 | - | 3 | 982 | |||
BC110620.1 | - | 3 | 1582 | 40035499 | ||
BC110621.1 | - | 3 | 1582 | 40035506 |