Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002126.4 | NP_002117.1 | 4 | 3662 | ||
XM_005257269.2 | XP_005257326.1 | 4 | 3645 | |||
XM_011524705.1 | XP_011523007.1 | 4 | 3119 | |||
XM_017024556.1 | XP_016880045.1 | 4 | 3116 | |||
GenBank mRNA | BC036093.1 | - | 4 | 3616 | 5288156 | |
M95585.1 | - | 4 | 3459 |