Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284194.1 | NP_001271123.1 | 3 | 2001 | ||
NM_173540.2 | NP_775811.2 | 3 | 1879 | |||
GenBank mRNA | BC036037.1 | - | 3 | 1879 | 5271548 |