Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNPNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198066.3 | NP_932332.1 | 6 | 1136 | ||
XM_005268012.2 | XP_005268069.1 | 7 | 1170 | |||
XM_006720238.3 | XP_006720301.1 | 6 | 1269 | |||
GenBank mRNA | AK001469.1 | - | 2 | 506 | ||
AK090577.1 | - | 6 | 1069 | |||
BC012179.1 | - | 6 | 938 | 4661046 |