Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DPYD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000110.3 | NP_000101.2 | 7, 8 | 895 | ||
XM_005270562.3 | XP_005270619.2 | 7, 8 | 895 | |||
XM_006710397.3 | XP_006710460.1 | 7, 8 | 895 | |||
XM_017000507.1 | XP_016855996.1 | 6, 7 | 778 | |||
XM_017000508.1 | XP_016855997.1 | 9, 10 | 1105 | |||
XM_017000509.1 | XP_016855998.1 | 8, 9 | 1003 | |||
XM_017000510.1 | XP_016855999.1 | 7, 8 | 892 | |||
GenBank mRNA | AB003063.1 | - | 7, 8 | 758 | ||
AK291217.1 | - | 7, 8 | 861 | |||
BC131777.1 | - | 7, 8 | 811 | 40123875 | ||
BC131778.1 | - | 7, 8 | 811 | 40123878 | ||
U09178.1 | - | 7, 8 | 839 | |||
U20938.1 | - | 7, 8 | 859 |