Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000414.3 | NP_000405.1 | 3, 4 | 346 | ||
NM_001199291.2 | NP_001186220.1 | 4, 5 | 545 | |||
NM_001199292.1 | NP_001186221.1 | 2, 3 | 292 | |||
NM_001292027.1 | NP_001278956.1 | 4, 5 | 411 | |||
NM_001292028.1 | NP_001278957.1 | 3, 4 | 480 | |||
GenBank mRNA | AK094049.1 | - | 3, 4 | 224 | ||
AK293254.1 | - | 3, 4 | 275 | |||
AK298075.1 | - | 4, 5 | 473 | |||
AK299685.1 | - | 4, 5 | 316 | |||
AK301212.1 | - | 2, 3 | 238 | |||
AK312449.1 | - | 3, 4 | 292 | |||
BC003098.1 | - | 3, 4 | 280 | 3502915 | ||
DA296575.1 | - | 3, 4 | 316 | |||
DA426062.1 | - | 3, 4 | 336 | |||
DA508630.1 | - | 3, 4 | 347 | |||
DC325873.1 | - | 4, 5 | 535 | |||
KU178135.1 | - | 3, 4 | 213 | |||
KU178136.1 | - | 3, 4 | 401 | |||
X87176.1 | - | 3, 4 | 261 |