Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNA21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002175.2 | NP_002166.2 | 1 | 769 | ||
GenBank mRNA | V00540.1 | - | 1 | 728 |
Gene Symbol: IFNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | E00175.1 | - | 1 | 705 |