Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFG3L1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003226.1 | 11 | 2731 | |||
NR_003227.1 | 10 | 2492 | ||||
GenBank mRNA | AK056488.1 | - | 12 | 2337 | ||
AK090856.1 | - | 2 | 1942 | |||
AK091293.1 | - | 11 | 2694 | |||
AK097044.1 | - | 10 | 2479 | |||
AK126911.1 | - | 9 | 2522 | |||
AK128352.1 | - | 4 | 2331 | |||
AK128804.1 | - | 5 | 1858 | |||
AL832984.1 | - | 5 | 1955 | |||
BC021245.2 | - | 9 | 2304 |