Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256959.1 | NP_001243888.1 | 4 | 426 | ||
NM_001286.3 | NP_001277.1 | 5 | 492 | |||
NR_046428.1 | 5 | 492 | ||||
GenBank mRNA | AK289999.1 | - | 5 | 397 | ||
AK294764.1 | - | 4 | 281 | |||
BC117420.1 | - | 5 | 349 | 40125971 | ||
BC117424.1 | - | 5 | 349 | 40125975 | ||
D28475.2 | - | 5 | 377 | |||
DA229615.1 | - | 5 | 492 | |||
DA935587.1 | - | 5 | 438 | |||
X83378.1 | - | 5 | 351 | |||
X96391.1 | - | 5 | 325 | |||
X99473.1 | - | 5 | 325 | |||
X99474.1 | - | 5 | 325 | |||
X99475.1 | - | 5 | 325 |