Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001334.2 | NP_001325.1 | 5 | 709 | ||
GenBank mRNA | AK300112.1 | - | 3 | 332 | ||
AK308323.1 | - | 5 | 715 | |||
AK312354.1 | - | 5 | 674 | |||
BC049206.1 | - | 5 | 662 | 5189657 | ||
X77383.1 | - | 5 | 709 |