Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ADAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278113.1 | NP_001265042.1 | 9 | 905 | ||
NM_001278114.1 | NP_001265043.1 | 10 | 962 | |||
NM_001464.4 | NP_001455.3 | 10 | 962 | |||
XM_005273468.1 | XP_005273525.1 | 10 | 900 | |||
GenBank mRNA | AJ133005.1 | - | 10 | 862 | ||
AK301734.1 | - | 10 | 900 | |||
BC034957.2 | - | 9 | 825 | 4826530 | ||
U38805.1 | - | 10 | 926 | |||
X99374.1 | - | 10 | 858 |