Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHROOM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001649.3 | NP_001640.1 | 4 | 596 | ||
XM_005274500.4 | XP_005274557.2 | 4 | 646 | |||
GenBank mRNA | BC140866.1 | - | 4 | 557 | 9021734 | |
X83543.1 | - | 4 | 596 |