Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DLAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001931.4 | NP_001922.2 | 2 | 987 | ||
XM_011542647.2 | XP_011540949.1 | 2 | 634 | |||
GenBank mRNA | AK057299.1 | - | 2 | 987 | ||
AK223596.1 | - | 2 | 376 | |||
AK296274.1 | - | 2 | 266 | |||
AK297103.1 | - | 2 | 390 | |||
AK299562.1 | - | 2 | 205 | |||
AK303930.1 | - | 2 | 393 | |||
AL832703.1 | - | 2 | 359 | |||
BC039084.1 | - | 2 | 356 | 4508489 |