- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: ITGB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002213.4 | NP_002204.2 | 9 | 1486 | ||
XM_005247436.2 | XP_005247493.1 | 8 | 1629 | |||
XM_017006351.1 | XP_016861840.1 | 9 | 1257 | |||
XM_017006352.1 | XP_016861841.1 | 9 | 1196 | |||
XM_017006353.1 | XP_016861842.1 | 9 | 1228 | |||
XM_017006354.1 | XP_016861843.1 | 6 | 1371 | |||
XM_017006355.1 | XP_016861844.1 | 8 | 1106 | |||
GenBank mRNA | AK130168.1 | - | 2 | 111 | ||
AK315429.1 | - | 9 | 1222 | |||
BC006541.2 | - | 9 | 1478 | 2958666 | ||
J05633.1 | - | 9 | 1227 | |||
M35011.1 | - | 9 | 1210 | |||
X53002.1 | - | 9 | 1517 |