Gene Symbol: TRPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252020.1 | NP_001238949.1 | 4 | 733 | ||
NM_001252024.1 | NP_001238953.1 | 5 | 521 | |||
NM_002420.5 | NP_002411.3 | 4 | 435 | |||
GenBank mRNA | AB115498.1 | - | 1 | 92 | ||
AB115499.1 | - | 1 | 92 | |||
AB115500.1 | - | 1 | 92 | |||
AB115501.1 | - | 1 | 92 | |||
AB115502.1 | - | 1 | 92 | |||
AF071787.1 | - | 4 | 412 | |||
HM135790.1 | - | 5 | 516 | |||
HM135791.1 | - | 4 | 733 |