Gene Symbol: TRPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252020.1 | NP_001238949.1 | 8 | 1366 | ||
NM_001252024.1 | NP_001238953.1 | 9 | 1154 | |||
NM_002420.5 | NP_002411.3 | 8 | 1068 | |||
GenBank mRNA | AB115498.1 | - | 5 | 725 | ||
AB115499.1 | - | 5 | 725 | |||
AB115500.1 | - | 5 | 755 | |||
AB115501.1 | - | 5 | 725 | |||
AB115502.1 | - | 5 | 707 | |||
AF071787.1 | - | 8 | 1045 | |||
HM135790.1 | - | 9 | 1149 | |||
HM135791.1 | - | 8 | 1366 |