Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002422.4 | NP_002413.1 | 1 | 237 | ||
GenBank mRNA | AK223283.1 | - | 1 | 155 | ||
AK223291.1 | - | 1 | 156 | |||
AK313310.1 | - | 1 | 153 | |||
BC069676.1 | - | 1 | 157 | 7262578 | ||
BC069716.1 | - | 1 | 157 | 7262590 | ||
BC074815.2 | - | 1 | 139 | 30915319 | ||
BC074869.2 | - | 1 | 139 | 30915468 | ||
BC105954.1 | - | 1 | 150 | 40033171 | ||
BC107490.1 | - | 1 | 150 | 40033168 | ||
BC107491.1 | - | 1 | 150 | 40033170 | ||
DA982602.1 | - | 1 | 237 | |||
DQ399598.1 | - | 1 | 58 | |||
J03209.1 | - | 1 | 94 | |||
X05232.1 | - | 1 | 137 |