Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002226.4 | NP_002217.3 | 26 | 5049 | ||
NM_145159.2 | NP_660142.1 | 25 | 4935 | |||
GenBank mRNA | AB209370.1 | - | 1 | 5378 | ||
AF020201.1 | - | 26 | 4692 | |||
BX105691.1 | - | 1 | 137 |