Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286457.1 | NP_001273386.1 | 20, 21 | 2284 | ||
NM_001286458.1 | NP_001273387.1 | 21, 22 | 2459 | |||
NM_001321858.1 | NP_001308787.1 | 20, 21 | 2210 | |||
NM_003470.2 | NP_003461.2 | 20, 21 | 2415 | |||
NR_135826.1 | 20, 21 | 2197 | ||||
XM_017023652.1 | XP_016879141.1 | 20, 21 | 2843 | |||
XM_017023653.1 | XP_016879142.1 | 20, 21 | 2279 | |||
GenBank mRNA | AK302481.1 | - | 21, 22 | 2385 | ||
AK302771.1 | - | 20, 21 | 2216 | |||
AK302912.1 | - | 21, 22 | 2403 | |||
AK316441.1 | - | 20, 21 | 2285 | |||
AY376241.1 | - | 20, 21 | 2246 | |||
Z72499.1 | - | 20, 21 | 2415 |