Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003635.3 | NP_003626.1 | 3 | 1065 | ||
XM_005270255.3 | XP_005270312.1 | 3 | 1065 | |||
XM_011540310.2 | XP_011538612.1 | 2 | 1634 | |||
XM_017016857.1 | XP_016872346.1 | 3 | 1065 | |||
GenBank mRNA | AB208870.1 | - | 1 | 222 | ||
AK303648.1 | - | 4 | 863 | |||
BC035711.1 | - | 3 | 818 | 5587622 | ||
BC110588.1 | - | 1 | 297 | 40011898 | ||
BC110589.1 | - | 1 | 297 | 40012703 | ||
U36601.1 | - | 1 | 288 |