Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322019.1 | NP_001308948.1 | 1, 2 | 178 | ||
NM_001322020.1 | NP_001308949.1 | 1, 2 | 178 | |||
NM_004544.3 | NP_004535.1 | 1, 2 | 178 | |||
NR_136155.1 | 1, 2 | 178 | ||||
NR_136156.1 | 1, 2 | 178 | ||||
NR_136158.1 | 1, 2 | 178 | ||||
XM_011511228.2 | XP_011509530.1 | 1, 2 | 165 | |||
GenBank mRNA | AB209440.1 | - | 1, 2 | 99 | ||
AF087661.1 | - | 1, 2 | 98 | |||
AF453834.1 | - | 1, 2 | 77 | |||
AK290778.1 | - | 1, 2 | 100 | |||
BC003417.1 | - | 1, 2 | 112 | 3451514 | ||
BC031332.1 | - | 1, 2 | 83 | 4872196 | ||
BI551914.1 | - | 162 |