Gene Symbol: NDUFC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204054.1 | NP_001190983.1 | 3 | 835 | ||
NM_001204055.1 | NP_001190984.1 | 3 | 973 | |||
NM_004549.5 | NP_004540.1 | 3 | 1042 | |||
GenBank mRNA | AF087899.1 | - | 3 | 1043 | ||
AF369951.1 | - | 3 | 649 | |||
AK023013.1 | - | 3 | 683 | |||
AK098173.1 | - | 3 | 457 | |||
BC007323.2 | - | 3 | 707 | 2990086 |