Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP9X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039590.2 | NP_001034679.2 | 10 | 1914 | ||
NM_001039591.2 | NP_001034680.2 | 10 | 1914 | |||
XM_005272675.4 | XP_005272732.1 | 10 | 2129 | |||
XM_005272676.4 | XP_005272733.1 | 10 | 2083 | |||
GenBank mRNA | AK294828.1 | - | 9 | 1315 | ||
X98296.1 | - | 9 | 1319 |