Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD11B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206741.1 | NP_001193670.1 | 7 | 1247 | ||
NM_005525.3 | NP_005516.1 | 6 | 1245 | |||
NM_181755.2 | NP_861420.1 | 7 | 1225 | |||
GenBank mRNA | BC012593.1 | - | 6 | 1194 | 4286028 |