Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MIPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005932.3 | NP_005923.2 | 5, 6 | 739 | ||
XM_011535097.2 | XP_011533399.1 | 5, 6 | 502 | |||
XM_011535098.2 | XP_011533400.1 | 5, 6 | 739 | |||
GenBank mRNA | AK291923.1 | - | 5, 6 | 615 | ||
BC009934.2 | - | 5, 6 | 711 | 4053687 | ||
U80034.1 | - | 5, 6 | 685 |