Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319116.1 | NP_001306045.1 | 6 | 675 | ||
NM_001319117.1 | NP_001306046.1 | 6 | 675 | |||
NM_001319118.1 | NP_001306047.1 | 6 | 603 | |||
NM_001319119.1 | NP_001306048.1 | 6 | 883 | |||
NM_001319120.1 | NP_001306049.1 | 6 | 700 | |||
NM_001319121.1 | NP_001306050.1 | 7 | 576 | |||
NM_001319122.1 | NP_001306051.1 | 6 | 668 | |||
NM_012100.3 | NP_036232.2 | 6 | 700 | |||
NR_134970.1 | 6 | 700 | ||||
GenBank mRNA | AF005050.2 | - | 6 | 675 | ||
AK001777.1 | - | 6 | 700 | |||
AK001977.1 | - | 6 | 672 | |||
AK021871.1 | - | 6 | 552 | |||
AK023005.1 | - | 6 | 614 | |||
AK301919.1 | - | 6 | 660 | |||
AK302809.1 | - | 6 | 667 | |||
AL519865.3 | - | 6 | 646 | |||
BC000653.2 | - | 6 | 582 | 3343505 | ||
BC004854.2 | - | 2 | 233 | |||
BI256589.1 | - | 6 | 566 | |||
BI821651.1 | - | 6 | 588 | |||
BM564135.1 | - | 6 | 663 | |||
BX403948.2 | - | 6 | 732 | |||
CN356162.1 | - | 6 | 594 |