Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS11E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014058.3 | NP_054777.2 | 7 | 727 | ||
XM_011531896.2 | XP_011530198.1 | 6 | 577 | |||
GenBank mRNA | AF064819.1 | - | 7 | 715 | ||
AK292787.1 | - | 7 | 713 | |||
AK313859.1 | - | 7 | 727 | |||
AY359017.1 | - | 7 | 694 | |||
BC113412.1 | - | 7 | 686 | 8322464 | ||
BC113414.1 | - | 7 | 686 | 8322466 |