Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014224.4 | NP_055039.1 | 4 | 407 | ||
GenBank mRNA | BC029055.1 | - | 4 | 386 | 5184316 | |
BC146999.1 | - | 4 | 407 | 9056927 | ||
BC171889.1 | - | 4 | 407 | 9054543 | ||
BC171897.1 | - | 4 | 407 | 9054551 |
Gene Symbol: PGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079807.3 | NP_001073275.1 | 4 | 408 | ||
GenBank mRNA | AK125628.1 | - | 4 | 422 | ||
AK225669.1 | - | 4 | 406 | |||
AK225674.1 | - | 4 | 434 | |||
AK225679.1 | - | 4 | 406 | |||
AK301286.1 | - | 4 | 408 | |||
AK301293.1 | - | 4 | 408 | |||
AK312941.1 | - | 4 | 408 | |||
BC171815.1 | - | 4 | 402 | 9054469 | ||
DA968889.1 | - | 4 | 426 |
Gene Symbol: PGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079808.3 | NP_001073276.1 | 4 | 408 | ||
GenBank mRNA | AK291864.1 | - | 4 | 406 | ||
BC150659.1 | - | 4 | 402 | 9057030 | ||
BC171808.1 | - | 4 | 402 | 9054462 | ||
BC171814.1 | - | 4 | 402 | 9054468 | ||
BC171910.1 | - | 4 | 402 | 9054564 | ||
BC171920.1 | - | 4 | 402 | 9054574 |