Gene Symbol: ADAMTS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014243.2 | NP_055058.2 | 3 | 191 | ||
XM_011532421.2 | XP_011530723.1 | 3 | 223 | |||
XM_011532422.2 | XP_011530724.1 | 3 | 722 | |||
GenBank mRNA | AB002364.1 | - | 3 | 143 | ||
AF247668.1 | - | 3 | 191 | |||
AK295131.1 | - | 3 | 111 | |||
BC130287.1 | - | 3 | 188 | 40146318 | ||
BC132735.1 | - | 3 | 188 | 40146757 |