Gene Symbol: ADAMTS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014243.2 | NP_055058.2 | 5 | 807 | ||
XM_011532421.2 | XP_011530723.1 | 5 | 839 | |||
XM_011532422.2 | XP_011530724.1 | 5 | 1338 | |||
GenBank mRNA | AB002364.1 | - | 5 | 759 | ||
BC130287.1 | - | 5 | 804 | 40146318 | ||
BC132735.1 | - | 5 | 804 | 40146757 |