Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015017.4 | NP_055832.3 | 6 | 797 | ||
NM_201624.2 | NP_963918.1 | 5 | 618 | |||
NM_201626.2 | NP_963920.1 | 6 | 797 | |||
XM_005270648.1 | XP_005270705.1 | 5 | 445 | |||
XM_005270649.1 | XP_005270706.1 | 5 | 445 | |||
XM_011541055.1 | XP_011539357.1 | 5 | 445 | |||
XM_011541056.2 | XP_011539358.1 | 5 | 445 | |||
XM_017000722.1 | XP_016856211.1 | 5 | 618 | |||
XM_017000723.1 | XP_016856212.1 | 5 | 569 | |||
XM_017000724.1 | XP_016856213.1 | 5 | 618 | |||
XM_017000725.1 | XP_016856214.1 | 5 | 618 | |||
XM_017000726.1 | XP_016856215.1 | 5 | 618 | |||
XM_017000727.1 | XP_016856216.1 | 5 | 461 | |||
XM_017000728.1 | XP_016856217.1 | 5 | 423 | |||
XM_017000729.1 | XP_016856218.1 | 5 | 445 | |||
GenBank mRNA | AB029020.1 | - | 5 | 539 | ||
AF383172.1 | - | 6 | 770 | |||
AF383173.1 | - | 5 | 591 | |||
AK022864.1 | - | 6 | 770 | |||
AK225499.1 | - | 3 | 238 | |||
AK308402.1 | - | 5 | 509 | |||
BC016663.1 | - | 6 | 651 | 3874822 |