Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ERAP1 SIRNA Assays ›
Gene Symbol:
ERAP1
Gene Name:
endoplasmic reticulum aminopeptidase 1
Gene Aliases:
A-LAP, ALAP, APPILS, ARTS-1, ARTS1, ERAAP, ERAAP1, PILS-AP, PILSAP
Chromosome Location:
Chr.5: 96759245 - 96935983 on Build GRCh38
UniGene ID:
Hs.666524
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
113526
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ERAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040458.1
NP_001035548.1
2
464
NM_001198541.1
NP_001185470.1
2
222
NM_016442.3
NP_057526.3
2
464
XM_005272015.4
XP_005272072.1
2
415
XM_005272016.4
XP_005272073.1
2
375
XM_011543480.2
XP_011541782.1
6
2684
XM_011543481.2
XP_011541783.1
6
2681
XM_011543484.2
XP_011541786.1
6
2680
XM_011543485.2
XP_011541787.1
5
2500
XM_011543486.2
XP_011541788.1
6
2684
XM_017009580.1
XP_016865069.1
7
2740
XM_017009581.1
XP_016865070.1
5
2526
XM_017009582.1
XP_016865071.1
2
199
XM_017009583.1
XP_016865072.1
2
410
GenBank mRNA
AB011097.2
-
1
138
AF106037.1
-
2
179
AF183569.1
-
2
184
AF222340.1
-
2
204
AK291636.1
-
2
214
AY028806.1
-
2
214
AY028807.1
-
2
214
AY358590.1
-
2
202
BC030775.1
-
2
216
4824919
DA327238.1
-
2
465
DA704693.1
-
2
518
JX962699.1
-
1
117
JX962700.1
-
1
117
JX962701.1
-
1
117
JX962702.1
-
1
117
KM357870.1
-
1
117
KM357871.1
-
1
117
KM357872.1
-
1
117
KM357873.1
-
1
117
KM357874.1
-
1
117
KM357875.1
-
1
117
KM357876.1
-
1
117
KM357877.1
-
1
117
KM357880.1
-
1
117
KM357881.1
-
1
117
KM357882.1
-
1
117
KM357883.1
-
1
117
KM357884.1
-
1
117
KM357885.1
-
1
117
KM357886.1
-
1
117
KM357887.1
-
1
117
KM357889.1
-
1
117
KM357890.1
-
1
117
KM357891.1
-
1
117
Back To Top
More Information
Panther Classification:
Molecular Function -
metalloprotease
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
adaptive immune response
antigen processing and presentation of peptide antigen via MHC class I
membrane protein ectodomain proteolysis
regulation of blood pressure
response to bacterium
antigen processing and presentation of endogenous peptide antigen via MHC class I
peptide catabolic process
regulation of innate immune response
fat cell differentiation
positive regulation of angiogenesis
endopeptidase activity
aminopeptidase activity
interleukin-6 receptor binding
interleukin-1, Type II receptor binding
tumor necrosis factor receptor binding
protein binding
metalloexopeptidase activity
zinc ion binding
peptide binding
metalloaminopeptidase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0