Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPV6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018646.5 | NP_061116.5 | 15 | 2700 | ||
GenBank mRNA | AF365927.1 | - | 15 | 2687 | ||
AF365928.1 | - | 11 | 2114 | |||
AJ243500.2 | - | 15 | 2670 | |||
AJ243501.2 | - | 15 | 2670 | |||
AJ487964.2 | - | 16 | 2641 | |||
AK075103.1 | - | 9 | 2201 | |||
AK075206.1 | - | 5 | 1070 | |||
AY203947.1 | - | 12 | 2882 | |||
BC034814.1 | - | 1 | 342 | |||
KF534785.1 | - | 15 | 2699 | |||
U82613.1 | - | 1 | 522 | |||
X83877.1 | - | 1 | 522 |