Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CACNA1G SIRNA Assays ›
Gene Symbol:
CACNA1G
Gene Name:
calcium voltage-gated channel subunit alpha1 G
Gene Aliases:
Ca(V)T.1, Cav3.1, NBR13, SCA42
Chromosome Location:
Chr.17: 50560929 - 50629625 on Build GRCh38
UniGene ID:
Hs.591169
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
113585
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CACNA1G
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256324.1
NP_001243253.1
23
4706
NM_001256325.1
NP_001243254.1
23
4706
NM_001256326.1
NP_001243255.1
23
4706
NM_001256327.1
NP_001243256.1
23
4706
NM_001256328.1
NP_001243257.1
23
4706
NM_001256329.1
NP_001243258.1
23
4706
NM_001256330.1
NP_001243259.1
23
4706
NM_001256331.1
NP_001243260.1
23
4706
NM_001256332.1
NP_001243261.1
22
4637
NM_001256333.1
NP_001243262.1
23
4706
NM_001256334.1
NP_001243263.1
23
4706
NM_001256359.1
NP_001243288.1
23
4706
NM_001256360.1
NP_001243289.1
23
4706
NM_001256361.1
NP_001243290.1
23
4706
NM_018896.4
NP_061496.2
23
4706
NM_198376.2
NP_938190.1
22
4637
NM_198377.2
NP_938191.2
23
4706
NM_198378.2
NP_938192.1
23
4706
NM_198379.2
NP_938193.1
22
4637
NM_198380.2
NP_938194.1
23
4706
NM_198382.2
NP_938196.1
22
4637
NM_198383.2
NP_938197.1
22
4637
NM_198384.2
NP_938198.1
23
4706
NM_198385.2
NP_938199.1
23
4706
NM_198386.2
NP_938200.1
23
4706
NM_198387.2
NP_938201.1
22
4637
NM_198388.2
NP_938202.1
22
4637
NM_198396.2
NP_938406.1
22
4637
NR_046054.1
23
4706
NR_046055.1
23
4706
NR_046056.1
23
4706
NR_046057.1
22
4637
NR_046058.1
22
4637
XM_006722160.3
XP_006722223.1
22
4776
XM_006722161.3
XP_006722224.1
22
4725
GenBank mRNA
AB012043.1
-
23
4342
AB032949.2
-
16
2668
AF126965.1
-
22
4420
AF126966.1
-
22
4420
AF134985.1
-
22
4245
AF134986.1
-
23
4314
AF190860.1
-
22
4461
AF227744.1
-
23
4314
AF227745.1
-
23
4314
AF227746.1
-
22
4245
AF227747.1
-
22
4245
AF227748.1
-
22
4245
AF227749.1
-
23
4314
AF227750.1
-
23
4314
AF227751.1
-
23
4314
AK296513.1
-
10
1417
BC110995.1
-
22
4637
5502027
DQ494449.1
-
22
4245
DQ494450.1
-
22
4245
DQ494451.1
-
22
4245
DQ494452.1
-
22
4245
DQ494453.1
-
22
4245
DQ494454.1
-
22
4245
DQ494455.1
-
22
4245
DQ494456.1
-
22
4245
DQ494457.1
-
23
4314
DQ494458.1
-
23
4314
DQ494459.1
-
23
4314
DQ494460.1
-
23
4314
DQ494461.1
-
23
4314
DQ494462.1
-
23
4314
DQ494463.1
-
23
4314
DQ494464.1
-
23
4314
DQ494465.1
-
23
4314
DQ494466.1
-
23
4314
DQ494467.1
-
23
4314
DQ494468.1
-
23
4314
DQ494469.1
-
23
4314
DQ494470.1
-
23
4314
DQ494471.1
-
23
4314
DQ494472.1
-
22
4245
DQ494473.1
-
23
4314
DQ494474.1
-
23
4314
DQ494475.1
-
23
4314
DQ494476.1
-
23
4314
DQ494477.1
-
23
4314
DQ494478.1
-
23
4314
DQ494479.1
-
23
4314
Back To Top
More Information
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
response to nickel cation
regulation of ion transmembrane transport
regulation of membrane potential
positive regulation of calcium ion-dependent exocytosis
regulation of atrial cardiac muscle cell membrane depolarization
calcium ion import
calcium ion transmembrane transport
membrane depolarization during action potential
SA node cell action potential
AV node cell action potential
SA node cell to atrial cardiac muscle cell signalling
AV node cell to bundle of His cell signaling
membrane depolarization during AV node cell action potential
membrane depolarization during SA node cell action potential
regulation of heart rate by cardiac conduction
low voltage-gated calcium channel activity
voltage-gated calcium channel activity involved in AV node cell action potential
voltage-gated calcium channel activity involved SA node cell action potential
scaffold protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0