Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021137.4 | NP_066960.1 | 3 | 702 | ||
XM_017024993.1 | XP_016880482.1 | 3 | 658 | |||
GenBank mRNA | AK300584.1 | - | 2 | 144 | ||
AK312387.1 | - | 3 | 463 | |||
BC001643.2 | - | 3 | 449 | 2823154 | ||
BC001949.1 | - | 3 | 449 | 2823154 | ||
BC003694.2 | - | 3 | 464 | |||
BT020121.1 | - | 2 | 253 | |||
CR456819.1 | - | 2 | 253 | |||
M80783.1 | - | 3 | 406 |