Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
綜合式 CRO CDMO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
企業服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADAM22 SIRNA Assays ›
Gene Symbol:
ADAM22
Gene Name:
ADAM metallopeptidase domain 22
Gene Aliases:
ADAM 22, MDC2
Chromosome Location:
Chr.7: 87934143 - 88202889 on Build GRCh38
UniGene ID:
Hs.256398
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
113658
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADAM22
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001324417.1
NP_001311346.1
8
979
NM_001324418.1
NP_001311347.1
8
982
NM_001324419.1
NP_001311348.1
8
979
NM_001324420.1
NP_001311349.1
8
982
NM_001324421.1
NP_001311350.1
8
982
NM_004194.4
NP_004185.1
8
982
NM_016351.5
NP_057435.2
8
982
NM_021721.4
NP_068367.1
8
982
NM_021722.5
NP_068368.2
8
982
NM_021723.4
NP_068369.1
8
982
XM_005250445.3
XP_005250502.1
7
830
XM_006716028.3
XP_006716091.1
7
830
XM_006716029.3
XP_006716092.1
7
830
XM_011516318.1
XP_011514620.1
8
874
XM_011516319.2
XP_011514621.1
8
958
XM_011516320.2
XP_011514622.1
7
830
XM_011516321.2
XP_011514623.1
7
830
XM_011516322.1
XP_011514624.1
8
874
XM_011516323.1
XP_011514625.1
8
874
XM_011516324.1
XP_011514626.1
8
546
XM_017012329.1
XP_016867818.1
9
925
XM_017012330.1
XP_016867819.1
9
925
XM_017012331.1
XP_016867820.1
8
955
XM_017012332.1
XP_016867821.1
8
874
XM_017012333.1
XP_016867822.1
8
874
XM_017012334.1
XP_016867823.1
9
925
XM_017012335.1
XP_016867824.1
9
925
XM_017012336.1
XP_016867825.1
8
874
XM_017012337.1
XP_016867826.1
9
925
XM_017012338.1
XP_016867827.1
8
953
XM_017012339.1
XP_016867828.1
9
925
XM_017012341.1
XP_016867830.1
7
571
GenBank mRNA
AB009671.1
-
8
780
AF073291.2
-
8
705
AF155381.1
-
8
705
AF155382.1
-
8
705
AF158637.1
-
7
542
AK125432.1
-
7
845
AK291010.1
-
8
829
BC036029.1
-
8
874
BC062433.1
-
8
813
BC092465.1
-
8
874
BC125118.1
-
8
736
40118904
Back To Top
More Information
Panther Classification:
Molecular Function -
metalloprotease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
cell adhesion
negative regulation of cell adhesion
central nervous system development
adult locomotory behavior
myelination in peripheral nervous system
metalloendopeptidase activity
integrin binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qskjf:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0