Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324159.1 | NP_001311088.1 | 9 | 1064 | ||
NM_022355.3 | NP_071750.1 | 11 | 1557 | |||
NR_136706.1 | 12 | 2046 | ||||
XM_005256090.4 | XP_005256147.2 | 9 | 2378 | |||
XM_011523266.1 | XP_011521568.1 | 11 | 1539 | |||
XM_011523268.2 | XP_011521570.1 | 10 | 2529 | |||
XM_011523271.2 | XP_011521573.1 | 8 | 2293 | |||
XM_011523272.2 | XP_011521574.1 | 10 | 1510 | |||
XM_011523273.2 | XP_011521575.1 | 9 | 1202 | |||
XM_011523274.2 | XP_011521576.1 | 7 | 933 | |||
XM_017023541.1 | XP_016879030.1 | 9 | 2202 | |||
XM_017023542.1 | XP_016879031.1 | 8 | 2283 | |||
XM_017023543.1 | XP_016879032.1 | 10 | 1530 | |||
XM_017023544.1 | XP_016879033.1 | 10 | 1509 | |||
XM_017023545.1 | XP_016879034.1 | 9 | 1787 | |||
XM_017023546.1 | XP_016879035.1 | 8 | 2623 | |||
XM_017023547.1 | XP_016879036.1 | 6 | 695 | |||
GenBank mRNA | AJ295149.1 | - | 11 | 1492 | ||
AK092884.1 | - | 12 | 2046 | |||
AK315090.1 | - | 10 | 1534 | |||
AY358454.1 | - | 10 | 1318 | |||
BC024021.1 | - | 10 | 1280 | 4616323 |