Search Thermo Fisher Scientific
Gene Symbol: ADAM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033274.4 | NP_150377.1 | 4 | 416 | ||
XM_005266003.2 | XP_005266060.1 | 4 | 368 | |||
GenBank mRNA | AF311317.1 | - | 4 | 325 | ||
AF326918.1 | - | 4 | 337 | |||
AK091464.1 | - | 4 | 368 | |||
AK308182.1 | - | 4 | 368 | |||
Y13786.2 | - | 4 | 366 |