Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MMP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032278.2 | NP_001027449.1 | 3 | 682 | ||
NM_024302.4 | NP_077278.1 | 3 | 682 | |||
NM_032950.3 | NP_116568.1 | 3 | 682 | |||
NR_111988.1 | 3 | 682 | ||||
XM_011525225.1 | XP_011523527.1 | 3 | 682 | |||
XM_011525226.2 | XP_011523528.1 | 4 | 794 | |||
XM_011525227.1 | XP_011523529.1 | 4 | 423 | |||
XM_011525228.1 | XP_011523530.1 | 4 | 448 | |||
XM_011525229.2 | XP_011523531.1 | 3 | 323 | |||
XM_011525230.1 | XP_011523532.1 | 3 | 682 | |||
XM_011525231.1 | XP_011523533.1 | 3 | 682 | |||
XM_011525232.2 | XP_011523534.2 | 3 | 682 | |||
XM_017025061.1 | XP_016880550.1 | 4 | 490 | |||
XM_017025062.1 | XP_016880551.1 | 3 | 682 | |||
XM_017025063.1 | XP_016880552.1 | 3 | 682 | |||
XM_017025064.1 | XP_016880553.1 | 3 | 682 | |||
GenBank mRNA | AF219624.1 | - | 3 | 521 | ||
AF315683.1 | - | 3 | 367 | |||
AF330002.1 | - | 3 | 627 | |||
AK122604.1 | - | 3 | 682 | |||
AY358987.1 | - | 3 | 557 | |||
BC002631.2 | - | 3 | 599 | 3610296 | ||
BC011774.2 | - | 3 | 522 |