Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543246.2 | XP_011541548.1 | 3 | 432 | ||
XM_011543247.2 | XP_011541549.1 | 3 | 397 | |||
XM_011543248.2 | XP_011541550.1 | 3 | 517 | |||
XM_017009174.1 | XP_016864663.1 | 2 | 1191 | |||
XM_017009176.1 | XP_016864665.1 | 3 | 397 | |||
GenBank mRNA | AJ311904.1 | - | 4 | 1044 | ||
AK128729.1 | - | 3 | 397 |