Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIBADH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152740.3 | NP_689953.1 | 3 | 535 | ||
XM_017011719.1 | XP_016867208.1 | 4 | 565 | |||
GenBank mRNA | AB050000.1 | - | 3 | 409 | ||
AF529362.1 | - | 3 | 322 | |||
AK025558.1 | - | 2 | 147 | |||
AK316605.1 | - | 3 | 340 | |||
BC020167.1 | - | 3 | 396 | |||
BC032324.1 | - | 3 | 420 | 3890576 | ||
BP343005.1 | - | 3 | 535 | |||
BQ051350.1 | - | 517 |