Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPXM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198148.2 | NP_937791.2 | 10 | 1728 | ||
XM_005269528.3 | XP_005269585.1 | 10 | 1578 | |||
XM_011539283.2 | XP_011537585.1 | 10 | 1578 | |||
XM_011539285.2 | XP_011537587.1 | 10 | 1360 | |||
XM_011539286.1 | XP_011537588.1 | 9 | 1302 | |||
XM_017015673.1 | XP_016871162.1 | 11 | 1329 | |||
XM_017015674.1 | XP_016871163.1 | 4 | 504 | |||
GenBank mRNA | AK296365.1 | - | 10 | 1135 | ||
AK300506.1 | - | 6 | 689 | |||
AK304814.1 | - | 10 | 1539 | |||
AL833732.1 | - | 8 | 906 | |||
AY358565.1 | - | 10 | 1728 | |||
BC027623.1 | - | 12 | 1451 | |||
BC036789.1 | - | 10 | 1558 | 5752707 | ||
BC137492.1 | - | 10 | 1723 | 9021492 | ||
BC146862.1 | - | 10 | 1723 | 9056790 |