Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002425.2 | NP_002416.1 | 1 | 123 | ||
GenBank mRNA | AK222601.1 | - | 1 | 123 | ||
AK313960.1 | - | 1 | 123 | |||
BC002591.2 | - | 1 | 110 | 3140935 | ||
BT007442.1 | - | 1 | 86 | |||
X07820.1 | - | 1 | 108 |